EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Fragrance allergen substitution.
PMID 10750869
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W247006
Isoeugenyl acetate, ≥98%, FCC, FG
C12H14O3