EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Relationship of mu opioid receptor binding to activation of G-proteins in specific rat brain regions.
PMID 10751548
Abstract
This study investigated the relationship between mu receptor binding and mu agonist activation of G-proteins in the rat brain. To directly compare agonist potencies in receptor binding (K(i) values) and G-protein activation (K(s) values), both agonist-stimulated [(35)S]guanosine-5'-O-(gamma-thio)-triphosphate ([(35)S]GTPgammaS) and [(3)H]naloxone binding assays were conducted under identical conditions, using the full mu agonist [d-Ala(2), N-Me(4), Gly(5)-ol]-enkephalin (DAMGO). DAMGO exhibited biphasic competition of [(3)H]naloxone binding and stimulation of [(35)S]GTPgammaS binding in most regions. Whereas the high-affinity component represented a large percentage (50-80%) of total receptor sites, the high-affinity component of DAMGO-stimulated [(35)S]GTPgammaS binding was much lower, <30% of the total, and in most regions significant stimulation of [(35)S]GTPgammaS binding did not occur until the high-affinity binding sites were completely occupied. Moreover, the low-affinity potencies for DAMGO in receptor binding and G-protein activation were the same across different regions. Receptor-transducer amplification factors were calculated by the ratio of the apparent B(max) of net agonist-stimulated [(35)S]GTPgammaS binding to the B(max) of receptor binding. Amplification factors for the nine regions examined were relatively high and varied significantly across regions, from a ratio of 8 in the thalamus to 38 in the cortex, suggesting that the efficiency of mu opioid receptor coupling to G-proteins varies across brain regions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7384
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
C26H35N5O6