EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
High-affinity, non-peptide agonists for the ORL1 (orphanin FQ/nociceptin) receptor.
PMID 10753470
Abstract
The discovery of 8-(5,8-dichloro-1,2,3,4-tetrahydro-naphthalen-2-yl)-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one, 1a, as a high-affinity ligand for the human ORL1 (orphanin FQ/nociceptin) receptor led to the synthesis of a series of optimized ligands. These compounds exhibit high affinity for the human ORL1 receptor, exhibit moderate to good selectivity versus opioid receptors, and behave as full agonists in biochemical assays. In this paper we present the synthesis, structure-activity relationship (SAR), and biochemical characterization of substituted 1-phenyl-1,3,8-triazaspiro[4.5]decan-4-ones culminating in the discovery of 8-(5-methyl-1,2,3,4-tetrahydro-naphthalen-1-yl)-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one, 1p, and 8-acenaphten-1-yl-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one 1q, two high-affinity, potent ORL1 receptor agonists with good to moderate selectivity versus the other opioid receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML2133
Ro 65-6570 Hydrochloride, ≥98% (HPLC)
C25H25N3O · HCl