EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
The yvaJ gene of Bacillus subtilis encodes a 3'-to-5' exoribonuclease and is not essential in a strain lacking polynucleotide phosphorylase.
PMID 10762271
Abstract
Studies of Bacillus subtilis RNases that are involved in mRNA degradation reveal a different pattern from that of Escherichia coli. A strain lacking polynucleotide phosphorylase, the major 3'-to-5' exoribonuclease activity in cell extracts, is viable. Here, we show that the B. subtilis yvaJ gene encodes a second 3'-to-5' exoribonuclease. A strain lacking both of these RNases grows slowly but is viable. The existence of another, as yet unknown, 3'-to-5' exoribonuclease in B. subtilis is suggested.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
DCL6B100 DEAE–Sepharose®, CL-6B