EMAIL THIS PAGE TO A FRIEND
Arthritis and rheumatism
Interleukin-1 induction of collagenase 3 (matrix metalloproteinase 13) gene expression in chondrocytes requires p38, c-Jun N-terminal kinase, and nuclear factor kappaB: differential regulation of collagenase 1 and collagenase 3.
PMID 10765924
Abstract
To examine the mechanism of interleukin-1 (IL-1)-induced collagenase 3 (matrix metalloproteinase 13 [MMP-13]) gene expression in cultured chondrocytes for the purpose of better understanding how the gene is induced in these cells, and how it contributes to cartilage degradation in osteoarthritis. The transcriptional and posttranscriptional responses of the MMP-13 gene to IL-1 were assessed first. Then, direct inhibitors of mitogen-activated protein kinase (MAPK) signaling pathways and a constitutive repressor of nuclear factor kappaB (NF-kappaB) were used to assess the role of each pathway in IL-1-mediated induction of MMP-13. We found that IL-1 induction of MMP-13 requires p38 activity, c-Jun N-terminal kinase (JNK) activity and NF-kappaB translocation. These results suggest that both NF-kappaB and activator protein 1 transcription factors are necessary for IL-1 induction of MMP-13. We also compared the signaling pathways necessary for IL-1 to stimulate collagenase 1 (MMP-1) in articular chondrocytes and chondrosarcoma cells and found that IL-1 induction of MMP-1 requires different pathways from those required by MMP-13. In chondrosarcoma cells, MMP-1 induction depends on p38 and MEK (an MAPK kinase of the extracellular signal-regulated kinase pathway) and does not require JNK or NF-kappaB. In articular chondrocytes, inhibition of MEK had no effect, while inhibition of p38 gave variable results. These studies demonstrate, for the first time, that p38, JNK, and NF-kappaB are required for IL-1 induction of MMP-13. The results also highlight the differential requirements for signaling pathways in the induction of MMP-1 and MMP-13. Additionally, they demonstrate that induction of MMP-1 by IL-1 in chondrocytic cells depends on unique combinations of signaling pathways that are cell type-specific.