EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Asymmetric reduction of ethyl 2-methyl e-oxobutanoate by fungi.
PMID 10766507
Abstract
Seven fungi, which are found to reduce ethyl 3-oxobutanoate in high yields, were tested for their reducing ability for ethyl 2-methyl 3-oxobutanoate. We obtained some interesting findings. In particular, Penicillium purpurogenum reduced ethyl 2-methyl 3-oxobutanoate to the corresponding alcohols with the diastereomer (anti/syn) ratio of 93/7 with the enantiomeric excess of anti-(2S,3S)- and syn-(2S,3R)- hydroxy esters of 90 and >99 ee%, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E35400
Ethyl 2-methylacetoacetate, 90%
C7H12O3