EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
LC analysis of hepatoprotective diterpenoids from Andrographis paniculata.
PMID 10768362
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
49879
Neoandrographolide, analytical standard
C26H40O8