EMAIL THIS PAGE TO A FRIEND
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme
[Development of donepezil hydrochloride (Aricept) for the treatment of Alzheimer's disease].
PMID 10771671
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1224981 Donepezil hydrochloride, United States Pharmacopeia (USP) Reference Standard
C24H29NO3 · HCl