EMAIL THIS PAGE TO A FRIEND
Biulleten' eksperimental'noi biologii i meditsiny
[Disturbed functioning of enzyme systems of microsomal oxidation, glucuro-, and glutathione conjugation of xenobiotics in the liver of rats intoxicated by deoxycholate and their correction].
PMID 10776570
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D98807
N,N-Diethylnicotinamide, 99%
C10H14N2O