EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
A method for the rapid preparation of 5-vinyluracil in high yield.
PMID 10793664
Abstract
A method for the rapid preparation of the thymineanalogue, 5-vinyluracil, in 83% yield from 5-(1-hydroxyethyl)uracil via the methanesulphonyl ester is reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
570656
5-Vinyluracil, 95%
C6H6N2O2