EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Improvement by low doses of nociceptin on scopolamine-induced impairment of learning and/or memory.
PMID 10794821
Abstract
The effects of fmol doses of nociceptin/orphanin FQ on scopolamine-induced impairment of learning and/or memory were examined using spontaneous alternation of Y-maze and step-down type passive avoidance tasks. While fmol doses of nociceptin alone had no effect on spontaneous alternation or passive avoidance behavior in normal mice, administration of nociceptin (10 and/or 100 fmol/mouse) 30 min before spontaneous alternation performance or the training session of the passive avoidance task, significantly improved the scopolamine-induced impairment of spontaneous alternation and passive avoidance behavior. This ameliorating effect was not antagonized by nocistatin (0.5 and 5.0 nmol/mouse, i.c.v.), naloxone benzoylhydrazone (2.3, 11.2, and 56.1 micromol/kg, s.c.) or nor-binaltorphimine (4.9 nmol/mouse, i.c.v.). These results indicated that very low doses of nociceptin ameliorate impairments of spontaneous alternation and passive avoidance induced by scopolamine, and suggested that this peptide has bidirectional modulatory effects on learning and memory; impairment at high doses and amelioration at low doses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N165
Naloxone benzoylhydrazone, solid
C26H27N3O4