EMAIL THIS PAGE TO A FRIEND
Science & justice : journal of the Forensic Science Society
A method for the rapid detection of the zopiclone degradation product 2-amino-5-chloropyridine.
PMID 10795415
Abstract
Zopiclone (Zimovane) is a cyclopyrrolone compound which exhibits hypnotic and sedative effects while also exhibiting anticonvulsant and muscle relaxant activities. The detection and quantification of zopiclone is difficult. It has a high molecular weight compared to most other commonly used drugs, therapeutic levels are not high, and it is unstable in nucleophilic solvents. A degradation product of zopiclone, 2-amino-5-chloropyridine (ACP) together with a method for its detection using high-performance liquid chromatography with diode array detection has been described previously. An account is presented of a simple method for the detection of ACP using gas chromatography with mass selective detection (GC/MS) which will facilitate detection of zopiclone use as part of a routine screen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A46803
2-Amino-5-chloropyridine, 98%
C5H5ClN2