EMAIL THIS PAGE TO A FRIEND
Trends in biotechnology
The synthesis of chiral cyanohydrins by oxynitrilases.
PMID 10802560
Abstract
Enantiomerically pure cyanohydrins are important synthetic intermediates for pharmaceuticals and agrochemicals. They are produced by enzyme-catalysed synthesis using oxynitrilases. Sufficient quantities of enzyme are available via cheap natural sources and there have been recent advances in overexpression production of cyanohydrins on an industrial scale.
Related Materials