EMAIL THIS PAGE TO A FRIEND
Nature medicine
Caspase 8 is deleted or silenced preferentially in childhood neuroblastomas with amplification of MYCN.
PMID 10802708
Abstract
Caspase 8 is a cysteine protease regulated in both a death-receptor-dependent and -independent manner during apoptosis. Here, we report that the gene for caspase 8 is frequently inactivated in neuroblastoma, a childhood tumor of the peripheral nervous system. The gene is silenced through DNA methylation as well as through gene deletion. Complete inactivation of CASP8 occurred almost exclusively in neuroblastomas with amplification of the oncogene MYCN. Caspase 8-null neuroblastoma cells were resistant to death receptor- and doxorubicin-mediated apoptosis, deficits that were corrected by programmed expression of the enzyme. Thus, caspase 8 acts as a tumor suppressor in neuroblastomas with amplification of MYCN.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV32030
Anti-MyCN antibody produced in rabbit, IgG fraction of antiserum