EMAIL THIS PAGE TO A FRIEND
Pharmacogenetics
Can caffeine metabolism be used as an in-vivo probe for human flavin-containing monooxygenase activity?
PMID 10803685
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC440
Benzydamine N-oxide hydrogen maleate
C19H23N3O2·C4H4O4