EMAIL THIS PAGE TO A FRIEND
Organic letters
Supramolecular phthalocyanine dimers based on the secondary dialkylammonium cation/dibenzo-24-crown-8 recognition motif
PMID 10804553
Abstract
[formula: see text] New unsymmetrically substituted DB24C8-phthalocyanines, which are able to form complexes with suitable dialkylammonium cations, have been prepared. These complexes most probably have a pseudorotaxane geometry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
459607
4,5-Dioctyloxy-1,2-benzenedicarbonitrile, 97%
C24H36N2O2