EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Raman excitation profiles and excited state molecular configurations of three isomeric phenyl pyridines.
PMID 10809061
Abstract
Contributions of different electronic states to Raman scattering have been studied by critical analyses of Raman excitation profiles (REPs) of several normal modes of vibration of three isomeric phenyl pyridines. In this context, possible structures and other interesting properties of the three molecules in the excited electronic states have been discussed. Normal mode characteristics are also described. Most likely a singlet state, lying in the vacuum ultraviolet region with respect to the ground state, is found to be playing a very significant role in the scattering phenomena.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
209732
3-Phenylpyridine, 97%
C11H9N
P33429
4-Phenylpyridine, 97%
C11H9N