EMAIL THIS PAGE TO A FRIEND
Journal of chromatographic science
Determination of selected herbicides and phenols in water and soils by solid-phase extraction and high-performance liquid chromatography.
PMID 10813519
Abstract
A high-performance liquid chromatography procedure or the determination of the herbicides simazine, propazine, bromacil, metoxuron, and hexazinone is elaborated. Stationary phases RP8 and RP18 and mixtures of methanol-water (2:1 and 1:1, v/v) as a mobile phase are applied for this purpose. The conditions for solid-phase extraction are established, allowing the separation of phenols and herbicides in their mixtures and the extraction of phenols (from river and coke plant water) and herbicides (from the soil samples).