EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
2-(trimethylsilyl)ethanethiol in nucleoside chemistry. A short route for preparing thionucleosides and their methyl disulfides.
PMID 10813923
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
364681
2-(Trimethylsilyl)ethanethiol, 95%
C5H14SSi