EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Scope and limitations of the Pd/BINAP-catalyzed amination of aryl bromides.
PMID 10814066
Abstract
Mixtures of Pd(2)(dba)(3) or Pd(OAc)(2) and BINAP catalyze the cross-coupling of amines with a variety of aryl bromides. Primary amines are arylated in high yield, and certain classes of secondary amines are also effectively transformed. The process tolerates the presence of several functional groups including methyl and ethyl esters, enolizable ketones, and nitro groups provided that cesium carbonate is employed as the base. Most reactions proceed to completion with 0.5-1.0 mol % of the palladium catalyst; in some cases, catalyst levels as low as 0.05 mol % Pd may be employed. Reactions are considerably faster if Pd(OAc)(2) is employed as the precatalyst, and the order in which reagents are added to the reaction has a substantial effect on reaction rate. It is likely that the catalytic process proceeds via bis(phosphine)palladium complexes as intermediates. These complexes are less prone to undergo undesirable side reactions which lead to diminished yields or catalyst deactivation than complexes of the corresponding monodentate triarylphosphines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219703
Hexylamine, 99%
C6H15N