EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Sequential acyl halide-aldehyde cyclocondensation and enzymatic resolution as a route to enantiomerically enriched beta-lactones.
PMID 10814079
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
135968
Acetyl bromide, 99%
C2H3BrO