EMAIL THIS PAGE TO A FRIEND
Organic letters
Enantioselective synthesis of the [6,6] spiroketal core of reveromycin A.
PMID 10814283
Abstract
[structure: see text] Reveromycin A (1) belongs to a family of microbial polyketides with unusual structural features and biological activities. The structure of 1 is composed of a [6,6] spiroketal core decorated with highly unsaturated side chains. As a prelude to the synthesis of 1, we present herein a short, efficient, and enantioselective synthesis of the C9-C21 fragment 5 (spiroketal core) of reveromycin A.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0654
Reveromycin A, solid
C36H52O11