EMAIL THIS PAGE TO A FRIEND
Journal of the American Society of Nephrology : JASN
Mitogen-activated protein kinase phosphorylation in kidneys of beta(s) sickle cell mice.
PMID 10820166
Abstract
Previous studies in beta(s) sickle cell mice demonstrated renal immunostaining for nitrotyrosine, which is putative evidence of peroxynitrite (ONOO(-)) formation. ONOO(-) is known to nitrate tyrosine residues of various enzymes, thereby interfering with phosphorylation and inactivating them. The present study examined the state of phosphorylation of mitogen-activated protein (MAP) kinase signal transduction enzymes, i.e., p38, c-Jun NH(2)-terminal kinase (JNK), and extracellular signal-regulated kinase (ERK). Western blot performed with antibodies directed against specific phosphorylated threonine/tyrosine residues of these enzymes demonstrated reduced phosphorylation of renal p38 and a trend toward reduced phosphorylation of ERK. In contrast, phosphorylation of renal JNK was markedly increased compared with normal mice. The abundance of MAP kinase phosphatase-1 (MKP-1), a key upstream enzyme that modulates phosphorylation of MAP kinases, was not different in beta(s) versus normal mice. To determine whether nitration of tyrosine by ONOO(-) was responsible for reduced phosphorylation of p38 and ERK, mercaptoethylguanidine (MEG), a compound known to reduce inducible isoform of nitric oxide synthase activity and to scavenge ONOO(-), was administered to beta(s) mice for 5 d. MEG was found to restore phosphorylation of p38 and ERK toward normal levels. These observations provide evidence that ONOO(-) (or closely related reaction products of NO) contributes to dephosphorylation of p38 and ERK, and presumably reduces activity of these enzymes. The increased phosphorylation of JNK, which suggests activation of this signaling pathway by extracellular stress signals, may play a role in apoptosis in the kidneys of these mice. The changes in phosphorylation of MAP kinase pathways found in this study could have important consequences for regulation of nuclear transcription factors, and thus renal function and pathology in sickle cell kidneys.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M9940
Mercaptoethylguanidine hemisulfate salt, ≥98%
C3H9N3S · 0.5H2SO4