EMAIL THIS PAGE TO A FRIEND
The Medical letter on drugs and therapeutics
New drugs for allergic conjunctivitis.
PMID 10825920
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0107
Pemirolast potassium, ≥98% (HPLC)
C10H7N6O·K