EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Induction of apoptosis in human papillomaviruspositive cancer cells by peptide aptamers targeting the viral E6 oncoprotein.
PMID 10829072
Abstract
Certain types of human papillomaviruses (HPVs) are closely linked to the development of human cancers. Herein, it is shown that intracellular targeting of the HPV16 E6 oncoprotein by E6-binding peptide aptamers resulted in the apoptotic elimination of HPV16-positive cancer cells, whereas HPV-negative cells were not affected. These results provide direct experimental evidence that the HPV E6 oncoprotein has antiapoptotic activity in HPV-positive tumor cells that is required for their survival. The E6-targeting molecules identified herein have implications for the development of therapeutic strategies for the treatment of HPV-associated dysplasias and cancers.