EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Glycosaminoglycan binding assays.
PMID 10840757
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0402 Heparin−Agarose, aqueous ethanol suspension