EMAIL THIS PAGE TO A FRIEND
Basic life sciences
Catechin and procyanidin levels in French wines: contribution to dietary intake.
PMID 10847859
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03910590
(±)-Catechin hydrate, primary reference standard
C15H14O6 · xH2O