EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Adenosine-dopamine interactions and ventilation mediated through carotid body chemoreceptors.
PMID 10849708
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A022
1,3-Dipropyl-8-(p-sulfophenyl)xanthine, powder
C17H20N4O5S