EMAIL THIS PAGE TO A FRIEND
Cancer biotherapy & radiopharmaceuticals
Rhenium-188--a generator-derived radioisotope for cancer therapy.
PMID 10851424
Abstract
Rhenium-188 (188Re) is an important therapeutic radioisotope which is obtained on demand as carrier-free sodium perrhenate by saline elution of the tungsten-188/rhenium-188 generator system. With a half-life of 16.9 hours and emission of a high energy beta particle (maximal energy of 2.12 MeV) and a gamma photon (155 keV, 15%) for imaging, 188Re can be provided at reasonable costs for routine preparation of radiopharmaceuticals for cancer treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
459984
Rhenium, foil, thickness 1.0 mm, 99.98% trace metals basis
Re
267317
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Re
204188
Rhenium, powder, 99.995% trace metals basis
Re
267279
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Re
GF57863931
Rhenium, wire reel, 0.1m, diameter 0.25mm, annealed, 99.97%
Re
GF60560513
Rhenium, wire reel, 0.1m, diameter 0.178mm, annealed, 99.97%
Re
GF90374441
Rhenium, wire reel, 0.1m, diameter 0.3mm, annealed, 99.97%
Re
GF77806367
Rhenium, wire reel, 0.1m, diameter 0.5mm, annealed, 99.97%
Re
GF63991929
Rhenium, wire reel, 0.1m, diameter 1.0mm, annealed, 99.97%
Re
GF14519156
Rhenium, wire reel, 0.2m, diameter 0.076mm, annealed, 99.97%
Re
GF59830139
Rhenium, wire reel, 0.2m, diameter 0.10mm, annealed, 99.97%
Re
GF96591875
Rhenium, wire reel, 0.2m, diameter 0.125mm, annealed, 99.97%
Re
GF02183217
Rhenium, wire reel, 0.2m, diameter 0.15mm, annealed, 99.97%
Re
GF70064658
Rhenium, wire reel, 0.2m, diameter 0.178mm, annealed, 99.97%
Re
GF32991486
Rhenium, wire reel, 0.2m, diameter 0.25mm, annealed, 99.97%
Re
GF48358435
Rhenium, wire reel, 0.2m, diameter 0.2mm, annealed, 99.97%
Re
GF96805884
Rhenium, wire reel, 0.2m, diameter 0.3mm, annealed, 99.97%
Re
GF25796010
Rhenium, wire reel, 0.2m, diameter 0.5mm, annealed, 99.97%
Re
GF98990050
Rhenium, wire reel, 0.2m, diameter 1.0mm, annealed, 99.97%
Re
GF14894163
Rhenium, wire reel, 0.5m, diameter 0.076mm, annealed, 99.97%
Re
GF78766745
Rhenium, wire reel, 0.5m, diameter 0.10mm, annealed, 99.97%
Re
GF73984797
Rhenium, wire reel, 0.5m, diameter 0.125mm, annealed, 99.97%
Re
GF72901324
Rhenium, wire reel, 0.5m, diameter 0.15mm, annealed, 99.97%
Re
GF58081945
Rhenium, wire reel, 0.5m, diameter 0.178mm, annealed, 99.97%
Re
GF23936104
Rhenium, wire reel, 0.5m, diameter 0.25mm, annealed, 99.97%
Re
GF44804325
Rhenium, wire reel, 0.5m, diameter 0.2mm, annealed, 99.97%
Re
GF00735353
Rhenium, wire reel, 0.5m, diameter 0.3mm, annealed, 99.97%
Re
GF24502115
Rhenium, wire reel, 0.5m, diameter 0.5mm, annealed, 99.97%
Re
GF61770225
Rhenium, wire reel, 1m, diameter 0.076mm, annealed, 99.97%
Re
GF45842733
Rhenium, wire reel, 1m, diameter 0.10mm, annealed, 99.97%
Re