EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Thermal tolerance of red shiner (Cyprinella lutrensis) after exposure to atrazine, terbufos, and their mixtures.
PMID 10857461
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45313
Terbufos, PESTANAL®, analytical standard
C9H21O2PS3