EMAIL THIS PAGE TO A FRIEND
Sbornik lekarsky
[Excitatory amino acid antagonists and potentiation of cortical evoked potentials].
PMID 10860126
Abstract
Changes of evoked potentials under the influence of NBQX (a non-NMDA receptor antagonist), MK-801 (a NMDA receptor antagonist) and GDEE (a nonselective antagonist of glutamate receptor) were studied. GDEE augmented potentiation and was marked progression of potentiation with increasing number of stimuli. There was no potentiation of responses in relation to the number of stimulus in a series in experiments with both MK-801 and NBQX. Interpretation of results with NBQX and MK-801 is difficult.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
309346
L-Glutamic acid diethyl ester hydrochloride, 97%
C9H17NO4 · HCl