EMAIL THIS PAGE TO A FRIEND
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Histamine and H2 agonist amthamine increase oxidative burst activity of human leukocytes determined by flow cytometry.
PMID 10864429
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4730
Amthamine dihydrobromide, ≥98% (HPLC), solid
C6H11N3S · 2HBr