EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A straightforward route to stereodefined functionalized cycloheptanols and cyclooctanols.
PMID 10864779
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C99000
Cycloheptanone, 99%
C7H12O
C109800
Cyclooctanone, 98%
C8H14O