EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Application of the BIRD sandwich for the rapid and accurate determination of (1)H-(1)H NMR coupling constants in higher order spin systems.
PMID 10869227
Abstract
A method is presented that allows for the convenient and reliable determination of (1)H-(1)H NMR coupling constants in higher order or symmetrically coupled spin systems. The method can be applied on any programmable FT-NMR spectrometer and is demonstrated here on micromole quantities of sample in a standard 5-mm NMR tube.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47930
Diethyl fumarate, purum, ≥98.0% (GC)
C8H12O4
D95654
Diethyl fumarate, 98%
C8H12O4