EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Intracisternal injection of somatostatin receptor 5-preferring agonists induces a vagal cholinergic stimulation of gastric emptying in rats.
PMID 10869415
Abstract
We previously showed that the somatostatin receptor 5 (sst(5))-preferring agonist BIM-23052 injected intracisternally (i.c. ; 0.8 nmol/rat) stimulated gastric emptying of a non-nutrient meal in conscious rats. In this study, we investigated the neural pathways and specificity of BIM-23052 action. BIM-23052 (0.4, 0.8, and 1.2 nmol/rat i.c.) stimulated gastric transit; values of gastric emptying were 65.5 +/- 6.5, 77.4 +/- 5.3, and 77.7 +/- 1.9%, respectively, compared with 43.2 +/-3.2% in i.c. saline group. Intravenous injection of BIM-23052 (0.8 nmol/rat) had no effect. BIM-23052 (0.8 nmol/rat i.c.) action was prevented by subdiaphragmatic vagotomy or atropine. Medullary thyrotropin-releasing hormone (TRH) is known to play a physiological role in the vagal stimulation of gastric motor function. TRH receptor antisense oligodeoxynucleotides injected i.c. with a regimen that prevented TRH (0.3 nmol/rat i.c.)-induced enhanced gastric emptying did not influence BIM-23052 stimulatory action. Somatostatin-28 (0.2-1.2 nmol/rat i.c.), which possesses a higher affinity than somatostatin-14 for sst(5), and the cyclic octapeptide des-AA(1,2,4,5,12,13)[D-Trp(8)]somatostatin (0.2-1.2 nmol/rat i.c.), an oligo-somatostatin analog that shares similar brain actions as somatostatin-28, induced a dose-related stimulation of gastric emptying. Somatostatin-14 and the preferring peptide agonists for sst(1), CH-275; sst(2), DC-32-87; sst(3), BIM-23056 and L-796,778; and sst(4), L-803,087 had no significant effect on gastric emptying when injected i.c. at 0.8 nmol/rat. These results show that BIM-23056 injected i.c. acts in the brain independently from medullary TRH to induce a vagal cholinergic stimulation of gastric emptying through the sst(5) receptor subtype.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S6135
Somatostatin 28, ≥97% (HPLC)
C137H207N41O39S3