EMAIL THIS PAGE TO A FRIEND
Japanese journal of pharmacology
Phenytoin and its metabolite, 5-(4-hydroxyphenyl)-5-phenylhydantoin, show bone resorption in cultured neonatal mouse calvaria.
PMID 10874595
Abstract
The effects of phenytoin and its major metabolite, 5-(4-hydroxyphenyl)-5-phenylhydantoin (HPPH), on bone resorption of neonatal mouse calvaria were examined in vitro. Both phenytoin and HPPH induced significant bone resorption as compared to the controls after 72 h in culture. This effect may be the cause of phenytoin-induced bone loss in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161543
5-(4-Hydroxyphenyl)-5-phenylhydantoin, 98%
C15H12N2O3