EMAIL THIS PAGE TO A FRIEND
Metabolism: clinical and experimental
Effect of triiodothyronine on reactive oxygen species generation by leukocytes, indices of oxidative damage, and antioxidant reserve.
PMID 10877210
Abstract
We have examined the effect of short-term triiodothyronine (T3) administration on reactive oxygen species (ROS) generation by leukocytes in 9 euthyroid subjects. At a dose of 60 microg/d orally for 7 days, T3 induced a significant increase in ROS generation by mononuclear cells (MNCs) from 183 +/- 102 mV at baseline to 313 +/- 111 mV on the seventh day (P < .02), and by polymorphonuclear leukocytes (PMNLs) from 195 +/- 94 mV at baseline to 302 +/- 104 mV on the seventh day (P < .02). There was also a significant increase in meta-tyrosine (P < .001) and ortho-tyrosine (P < .001), known indices of oxidative damage to proteins and amino acids. However, there was no increase in plasma thiobarbituric acid-reactive substances (TBARS), an index of oxidative damage to lipids, and in the level of carbonylated proteins, a less sensitive index to assess protein oxidation. There was no decrease in the level of antioxidants such as alpha-tocopherol, vitamin A, beta-carotene, lycopene, and lutein/zeaxanthin. The stimulatory effect on ROS generation may reflect a generalized increase in metabolic activity or may be a specific effect on NADPH oxidase in leukocyte membranes. The absence of a significant change in TBARS, carbonylated proteins, alpha-tocopherol, vitamin A, beta-carotene, lycopene, and lutein/zeaxanthin may reflect the short duration of the increased ROS load.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93851
DL-o-Tyrosine, ≥96.0% (NT)
C9H11NO3