EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Isolation and characterization of 2,3-dichloro-1-propanol-degrading rhizobia.
PMID 10877782
Abstract
2,3-Dichloro-1-propanol is more chemically stable than its isomer, 1, 3-dichloro-2-propanol, and is therefore more difficult to degrade. The isolation of bacteria capable of complete mineralization of 2, 3-dichloro-1-propanol was successful only from enrichments at high pH. The bacteria thus isolated were found to be members of the alpha division of the Proteobacteria in the Rhizobium subdivision, most likely Agrobacterium sp. They could utilize both dihaloalcohol substrates and 2-chloropropionic acid. The growth of these strains in the presence of 2,3-dichloro-1-propanol was strongly affected by the pH and buffer strength of the medium. Under certain conditions, a ladder of four active dehalogenase bands could be visualized from this strain in activity gels. The enzyme involved in the complete mineralization of 2,3-dichloro-1-propanol was shown to have a native molecular weight of 114,000 and consisted of four subunits of similar molecular weights.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36295
2,3-Dichloro-1-propanol, ≥97.0% (GC)
C3H6Cl2O