EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Modification of sorbitol MacConkey medium containing cefixime and tellurite for isolation of Escherichia coli O157:H7 from radish sprouts.
PMID 10877817
Abstract
A modified version of sorbitol MacConkey medium containing cefixime and tellurite (CT-SMAC medium) was produced by adding salicin and 4-methylumbelliferyl-beta-D-galactopyranoside to CT-SMAC medium; this medium was designated CT-SSMAC medium and was used to isolate Escherichia coli O157:H7 from radish sprouts. Of 101 non-E. coli bacteria isolated from radish sprouts that produced colorless colonies similar to colonies of E. coli O157:H7 grown on CT-SMAC medium, 92 (91%) formed colonies that were red to pink or were beta-galactosidase negative and colorless on CT-SSMAC medium. On the other hand, colonies of E. coli O157:H7 strains were colorless and beta-galactosidase positive on CT-SSMAC medium. Our results suggest that CT-SSMAC medium is more selective than CT-SMAC medium for isolating E. coli O157:H7.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
18588
Cefixime trihydrate, for microbiology, ≥98.0% (HPLC)
C16H15N5O7S2 · 3H2O