EMAIL THIS PAGE TO A FRIEND
Japanese journal of pharmacology
Effects of zaldaride maleate, an antidiarrheal compound, on intracellular Ca2+-mediated short-circuit current response in the rat colonic mucosa.
PMID 10887945
Abstract
The effect of zaldaride maleate (ZAL) on intracellular Ca2+-mediated short-circuit current (Isc) response in rat colonic mucosa was investigated. The Isc response to acetylcholine was partially reduced by the neuronal blocker tetrodotoxin. The tetrodotoxin-sensitive Isc response to acetylcholine was inhibited by ZAL. The tetrodotoxin-insensitive effect, which indicates direct stimulation of acetylcholine in colonic mucosa, was not affected by ZAL. The Isc response to the Ca2+ ionophore A23187 was reduced by ZAL in the presence of tetrodotoxin. These results suggest that ZAL reduces colonic ion secretion by inhibiting a tetrodotoxin-sensitive neuronal pathway and an intracellular Ca2+ pathway in epithelium.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1619
CGS-9343B, ≥98% (HPLC), powder
C26H28N4O2 · C4H4O4