EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Micelle-mediated extraction for concentrating hydrophobic organic compounds.
PMID 10893028
Abstract
An extraction method based on polymer-induced phase separation of aqueous micellar solutions of octyl-beta-D-thioglucoside (OTG) was assessed for concentrating hydrophobic analytes. Various hydrophobic compounds such as polycyclicaromatic hydrocarbons, alkylbenzenes, alkylphenols, chlorobenzenes, chlorophenols, phthalic esters, pesticides, and steroid hormones could be efficiently concentrated into a small volume of surfactant-rich phase, while hydrophilic matrix components remained in the bulk aqueous phase. The surfactant-rich phase containing concentrated OTG could be directly introduced into the hygro-organic mobile phase of high-performance liquid chromatography with ultra-violet photometric detection. The application of this method greatly enhanced the signal intensity in the chromatogram while reducing the interference of matrix components.
Related Materials