EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Identification of the Escherichia coli nicotinic acid mononucleotide adenylyltransferase gene.
PMID 10894752
Abstract
The gene (ybeN) coding for nicotinate mononucleotide adenylyltransferase, an NAD(P) biosynthetic enzyme, has been identified and overexpressed in Escherichia coli. This enzyme catalyzes the reversible adenylation of nicotinate mononucleotide and shows product inhibition. The rate of adenylation of nicotinate mononucleotide is at least 20 times faster than the rate of adenylation of nicotinamide mononucleotide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7764
Nicotinic acid mononucleotide
C11H14NO9P