EMAIL THIS PAGE TO A FRIEND
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society
Dissociation of parathyroid hormone and cyclic-3', 5'AMP effects on Na-Pi uptake by cells isolated from proximal straight tubules of rat kidney.
PMID 10898102
Abstract
Studies in respiratory alkalotic or short-term phosphate deprived rats raised the possibility that in straight portion of proximal tubules (PST) cAMP might be not a mediator of PTH in inhibition of phosphate reabsorption. The present experiments directly compared the sensitivity of Na-dependent phosphate [32P] (Na-Pi) uptake to PTH or cAMP by PCT or PST cells freshly prepared from outer cortex and outer stripe of outer medulla of rat kidney. The purity of the cells was examined by activity of enzymes specific for PST i.e. glutamine synthetase, gamma-glutamyl transpeptidase and creatine kinase, a marker enzyme for medullary thick ascending limb (MTAL) and distal convoluted tubule. Similar inhibition of Na-Pi uptake by 1-34 bPTH by PST and PCT cells was observed: -33.0 and -30.0% (ns), respectively. In contrast, dibutyryl cAMP decreased Na-Pi uptake only by PCT but not by PST cells: -31.0 and -3.6% (p<0.02), respectively. The 3-isobutyl-1-methylxanthine (IBMX), a phosphodiesterase inhibitor, resulted in slight stimulation of Na-Pi uptake by PST but strong inhibition by PCT cells: 7.8 vs -26.0% (p<0.001). In contrast to PCT in PST cells cAMP seems to play a minor role as a mediator of inhibition of Na-Pi uptake by PTH.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3671 Parathyroid Hormone Fragment 1-34 bovine, ≥97% (HPLC), powder
C183H288N54O50S2