EMAIL THIS PAGE TO A FRIEND
Current biology : CB
Rapid induction of apoptosis mediated by peptides that bind initiation factor eIF4E.
PMID 10898981
Abstract
Overexpression of the translation initiation factor eIF4E leads to cell transformation and occurs in a number of human cancers [1]. mRNA translation and cell growth can be regulated through the availability of eIF4E to form initiation complexes by binding to eIF4G. The availability of eIF4E is blocked through the binding of members of a family of eIF4E-binding proteins (4E-BPs) [2] [3]. Indeed, cell transformation caused by the overexpression of eIF4E can be reversed by the overexpression of 4E-BPs [4] [5] [6] [7] [8]. To study the role of eIF4E in cell transformation, we developed a series of peptides based on the conserved eIF4E-binding motifs in 4E-BPs and eIF4G [9] linked to the penetratin peptide-carrier sequence, which mediates the rapid transport of peptides across cell membranes. Surprisingly, introduction of these eIF4E-binding peptides into MRC5 cells led to rapid, dose-dependent cell death, with characteristics of apoptosis. Single alanine substitutions at key positions in the peptides impair their binding to eIF4E and markedly reduce their ability to induce apoptosis. A triple alanine substitution, which abolishes binding to eIF4E, renders the peptide unable to induce apoptosis. Our data provide strong evidence that the peptides induce apoptosis through binding to eIF4E. They do not induce apoptosis through inhibition of protein synthesis, as chemical inhibitors of translation did not induce apoptosis or affect peptide-induced cell death. Thus these new data indicate that eIF4E has a direct role in controlling cell survival that is not linked to its known role in mRNA translation.