EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Mechanism-based inactivation of cytochromes P450 2B1 and P450 2B6 by 2-phenyl-2-(1-piperidinyl)propane.
PMID 10901699
Abstract
2-Phenyl-2-(1-piperidinyl)propane (PPP), an analog of phencyclidine, was tested for its ability to inactivate cytochrome P450s (P450s) 2B1 and 2B6. PPP inactivated the 7-(benzyloxy)resorufin O-dealkylation activity of liver microsomes obtained from phenobarbital-induced rats with a K(I) of 11 microM. The 7-ethoxy-4-(trifluoromethyl)coumarin O-deethylation activity of purified rat liver P450 2B1 and expressed human P450 2B6 was inactivated by PPP in a reconstituted system containing NADPH-cytochrome P450 reductase and lipid. In the presence of NADPH, the loss of activity was time- and concentration-dependent, and followed pseudo first order kinetics. The rate of inactivation for P450 2B1 was 0.3 min(-1), and the concentration of PPP required to achieve half-maximal inactivation was 12 microM. The time for 50% of the P450 2B1 to become inactivated at saturating concentrations of PPP was 2.5 min. P450 2B6 was inactivated with a k(inact) of 0.07 min(-1), a K(I) of 1.2 microM, and a t(1/2) of 9.5 min. The inactivated P450s 2B1 and 2B6 lost about 25 and 15%, respectively, of their ability to form a CO-reduced complex, suggesting that the loss of activity was caused by a PPP modification of the apoprotein rather than the heme. The estimated partition ratio for P450s 2B1 and 2B6 with PPP was 31 and 15, respectively. The inactivation was not reversible and reductase activity was not affected. Coincubation of P450 2B1 and 2B6 with PPP and NADPH in the presence of an alternate substrate protected both enzymes from inactivation. The exogenous nucleophile GSH did not affect the rate of inactivation. PPP-inactivated P450s 2B1 and 2B6 were recognized on Western blots by an antibody generated to phencyclidine that had been conjugated to BSA. Stoichiometries of 1.4:1 and 0.7:1 were determined for the binding of a [3H]PPP metabolite to P450 2B1 and 2B6, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T2803
7-Ethoxy-4-(trifluoromethyl)coumarin, ≥98% (TLC)
C12H9F3O3