EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Occupational allergic contact dermatitis from 2,2-bis[4-(2-hydroxy-3-acryloxypropoxy)phenyl]-propane (epoxy diacrylate) in ultraviolet-cured inks.
PMID 10902601
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
411167
Bisphenol A glycerolate (1 glycerol/phenol) diacrylate, contains MEHQ as inhibitor
C27H32O8