EMAIL THIS PAGE TO A FRIEND
Lipids
Simultaneous derivatization of acyl and S-alkyl moieties of acyl thioesters by using trimethylsulfonium hydroxide for gas chromatographic analysis.
PMID 10907793
Abstract
The reaction of long-chain acyl thioesters, viz. lauric acid dodecyl thioester, 1,8-di-S-palmitoyl octanedithiol, and tristearoyl alpha-monothioglycerol, with trimethylsulfonium hydroxide in the presence of methanol leads to simultaneous derivatization of acyl moieties and S-alkyl moieties of acyl thioesters to the corresponding fatty acid methyl esters and methyl alkylsulfides, respectively. These derivatized products were analyzed by gas chromatographic technique.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T80489
Trimethylsulfonium iodide, 98%
C3H9IS