EMAIL THIS PAGE TO A FRIEND
DNA research : an international journal for rapid publication of reports on genes and genomes
The human homologue of flamingo, EGFL2, encodes a brain-expressed large cadherin-like protein with epidermal growth factor-like domains, and maps to chromosome 1p13.3-p21.1.
PMID 10907856