EMAIL THIS PAGE TO A FRIEND
Planta medica
Cytotoxic neolignans from the stem wood of Machilus obovatifolia.
PMID 10909257
Abstract
Four new cytotoxic neolignans, machilusol A (1), machilusol C (3), machilusol D (4), machilusol E (5) as well as two new inactive neolignans, machilusol B (2) and machilusol F (6), were isolated from the stem wood of Machilus obovatifolia. All structures were identified by means of spectroscopic analysis.