EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Solid phase synthesis of [18F]labelled peptides for positron emission tomography.
PMID 10915036
Abstract
A strategy for the solid phase synthesis of [18F]labelled peptides has been developed. The peptides were prepared on solid support and acylated with 4-[18F]fluorobenzoic acid using HATU within 3 min and the labelled peptide was released from the solid support within 7 min. The [18F]labelled peptides were produced in good purity with a specific activity of 20-25 GBq/micromol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
418846
4-Fluorobenzoic acid, 99%, purified by sublimation
C7H5FO2
128384
4-Fluorobenzoic acid, 98%
C7H5FO2